Tetraethylammonium acetate tetrahydrate, 96%
Tetraethylammonium acetate tetrahydrate, 96%
Tetraethylammonium acetate tetrahydrate, 96%
Thermo Scientific Chemicals

Tetraethylammonium acetate tetrahydrate, 96%

Phase transfer catalyst | CAS: 67533-12-4 | C10H23NO2·4H2O | 261.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 194945000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS67533-12-4
IUPAC Nametetraethylazanium acetate tetrahydrate
Molecular FormulaC10H31NO6
InChI KeyQNOAKQGNSWGYOE-UHFFFAOYSA-M
SMILESO.O.O.O.CC([O-])=O.CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO495 to 105 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only