Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Palladium(II) chloride, 59% Pd
Thermo Scientific Chemicals

Palladium(II) chloride, 59% Pd

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol
Have Questions?
Catalog NumberQuantityPackaging
19520025025 gGlass bottle
Catalog number 195200250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7647-10-1
IUPAC Namepalladium(2+) dichloride
Molecular FormulaCl2Pd
InChI KeyPIBWKRNGBLPSSY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Assay59 to 60 % (Pd)
Iron (Fe)=<15 ppm
Silver (Ag)=<30 ppm
Copper (Cu)=<15 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only