Forma™ Freezer Replacement Shelves
Actual product may vary
Forma™ Freezer Replacement Shelves
Forma™ Freezer Replacement Shelves
Forma™ Freezer Replacement Shelves
Forma™ Freezer Replacement Shelves
Thermo Scientific™
Forma™ Freezer Replacement Shelves
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
19533813.0 cu. ft. FreezersShelf, Behind-The-Mullion
19533917.3 cu. ft. FreezersShelf, Behind-The-Mullion
19534023.0 cu. ft. FreezersShelf, Behind-The-Mullion
22413413.0 cu. ft. FreezersShelf Kit
22413517.3 cu. ft. FreezersShelf Kit
22415823.0 cu. ft. FreezersShelf Kit
22418028 cu. ft. FreezersShelf Kit
Catalog number 195338
Price (USD)
-
For Use With (Equipment):
13.0 cu. ft. Freezers
Type:
Shelf, Behind-The-Mullion
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
  • Stainless steel
  • Kits include mounting hardware
Specifications
ColorSilver
For Use With (Equipment)13.0 cu. ft. Freezers
TypeShelf, Behind-The-Mullion
Capacity (English)13 cu. ft.
Capacity (Metric)368.1 L
StyleUpright
Unit SizeEach