Celestine Blue, pure
Celestine Blue, pure
Celestine Blue, pure
Thermo Scientific Chemicals

Celestine Blue, pure

CAS: 1562-90-9 | C17H18ClN3O4 | 363.798 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 195560100
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Chemical Identifiers
CAS1562-90-9
IUPAC Name{amino[7-(diethylamino)-3,4-dioxo-4,10-dihydro-3H-phenoxazin-1-yl]methylidene}oxidanium chloride
Molecular FormulaC17H18ClN3O4
InChI KeyREPMZEQSQQAHJR-UHFFFAOYSA-N
SMILES[Cl-].CCN(CC)C1=CC=C2NC3=C(OC2=C1)C(=O)C(=O)C=C3C(N)=[OH+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to dark brown to black
Appearance (Form)Powder
Infrared spectrumConforms
UV spectrumComplies
UV Lambda max638 to 648 nm (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only