D(+)-Raffinose pentahydrate, 99+%
D(+)-Raffinose pentahydrate, 99+%
D(+)-Raffinose pentahydrate, 99+%
Thermo Scientific Chemicals

D(+)-Raffinose pentahydrate, 99+%

CAS: 17629-30-0 | C18H32O16·5H2O | 594.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 195670250
Price (EUR)
96,65
Online exclusive
107,00
Save 10,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
96,65
Online exclusive
107,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS17629-30-0
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyMUPFEKGTMRGPLJ-ZQSKZDJDSA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sulfated ash=<0.1 % (on dry basis)
Specific optical rotation+103.0° to +107.0° (20°C, 589 nm) (c=10, water)
Solubility(10 % in water) Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only