Product Image
Applied Biosystems™
Eureka™ 1.0 Reagent Kit
Have Questions?
Catalog NumberQuantity
19585384 reactions
Catalog number 19585
Price (MXN)
-
Quantity:
384 reactions
Comprehensive Reagent Kit
The Eureka 1.0 Reagent Kit enables genotyping of SNPs and indels on Eureka pre-designed and custom genotyping panels. These kits are available for 384 reactions. The kits include reagents required to
• Produce next-generation sequencing (NGS)-ready libraries to be pooled
• Process samples on a NGS sequencer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Genotyping
Quantity384 reactions
Reagent TypeGenotyping reagents
Type1.0 Reagent Kit
Unit SizeEach