Boron standard solution, for AAS, 1 mg/ml B in water
Boron standard solution, for AAS, 1 mg/ml B in water
Boron standard solution, for AAS, 1 mg/ml B in water
Thermo Scientific Chemicals

Boron standard solution, for AAS, 1 mg/ml B in water

CAS: 7440-42-8 | B | 10.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 195865000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS7440-42-8
ChEBICHEBI:27560
Chemical Name or MaterialBoron standard solution
EINECS Number231-151-2
Formula Weight10.81
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only