Air Filter Replacement Kits for Ultra Low Temperature Freezers
Thermo Scientific™
Air Filter Replacement Kits for Ultra Low Temperature Freezers
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
195890Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series
195947Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers
195948Revco™ CxF, HERAfreeze™ HFC, TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers
AF34567HERAfreeze™ HFU-T, Revco™ UxF, Thermo Scientific™ TSU Series
AF34567LETSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE
332297H02TDE Series Chest Freezers
Catalog number 195890
Price (USD)
-
For Use With (Equipment):
Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filter
Specifications
DescriptionFor use with Forma 900 and 7000, HERAfreeze HFU B, Revco ExF/DxF, TSE/TSD Series of Upright ULT Freezers.
For Use With (Equipment)Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series
TypeFreezer Air Filter
StyleUpright
Unit SizeEach