Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3
Thermo Scientific Chemicals

Cobalt standard solution, for AAS, 1 mg/ml Co in 2 to 5% HNO3

CAS: 10141-05-6 | CoN2O6 | 182.941 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 195925000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS10141-05-6
CAS7732-18-5
ChEBICHEBI:86209
Chemical Name or MaterialCobalt standard solution
Formula Weight58.93
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only