Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3
Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3
Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3
Thermo Scientific Chemicals

Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 195935000
Price (USD)
75.80
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
75.80
Each
Specifications
CAS7440-50-8
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:30052
Chemical Name or MaterialCopper standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only