Air Filter Replacement Kits for Ultra Low Temperature Freezers
Thermo Scientific™
Air Filter Replacement Kits for Ultra Low Temperature Freezers
Easily replace the air filters on your ultra-low temperature freezer with these replacement parts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
195947Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers
195890Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series
195948Revco™ CxF, HERAfreeze™ HFC, TSC-Series 12.7, 17.0 and 20.0 cu. ft. Chest ULT Freezers
AF34567HERAfreeze™ HFU-T, Revco™ UxF, Thermo Scientific™ TSU Series
AF34567LETSX Series, Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE
332297H02TDE Series Chest Freezers
Catalog number 195947
Price (USD)
-
For Use With (Equipment):
Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers
Easily replace the air filters on your ultra-low temperature freezer with Thermo Scientific™ Air Filter Replacement Kits for Ultra Low Temperature Freezers.
Replacement air filter
Specifications
DescriptionFor use with Forma, HERAfreeze, Revco and TS-Series 3.0 cu. ft. Chest ULT Freezers
For Use With (Equipment)Revco™, TS-Series 3.0 cu. ft. Chest ULT Freezers
TypeFreezer Air Filter
Capacity (English)3 cu. ft.
Capacity (Metric)84.9 L
StyleChest
Unit SizeEach