Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3
Thermo Scientific Chemicals

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3

CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 196051000
Price (USD)
89.65
Online exclusive
99.90
Save 10.25
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
89.65
Online exclusive
99.90
Save 10.25
Each
Specifications
CAS7439-89-6
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:82664
Chemical Name or MaterialIron standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only