Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3
Thermo Scientific Chemicals

Lead standard solution, for AAS, 1 mg/ml Pb in 2-5% HNO3

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 196070025
Price (USD)
368.65
Online exclusive
409.00
Save 40.35
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
368.65
Online exclusive
409.00
Save 40.35
Each
Specifications
Boiling Point101.0°C
CAS10099-74-8
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:37187
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only