Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl
Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl
Thermo Scientific Chemicals

Lithium standard solution, for AAS, 1 mg/ml Li in 2% HCl

CAS: 7439-93-2 | Li | 6.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 196081000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS7439-93-2
CAS7647-01-0
CAS7732-18-5
ChEBICHEBI:30145
Chemical Name or MaterialLithium standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only