Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3
Thermo Scientific Chemicals

Magnesium standard solution, for AAS, 1 mg/ml Mg in 0.5N HNO3

CAS: 7439-95-4 | Mg | 24.30 g/mol
Have Questions?
Catalog NumberQuantity
196105000500 mL
Catalog number 196105000
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 28-Dec-2026
135.00
Save 20.35
Each
Specifications
CAS7439-95-4
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:25107
Chemical Name or MaterialMagnesium standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only