Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3
Thermo Scientific Chemicals

Mercury standard solution, for AAS, 1 mg/ml Hg in 2 to 5% HNO3

CAS: 7783-34-8 | H2HgN2O7 | 342.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 196125000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7783-34-8
IUPAC Namemercury(2+) hydrate dinitrate
Molecular FormulaH2HgN2O7
InChI KeyKVICROHOONHSRH-UHFFFAOYSA-N
SMILESO.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Strength990 to 1010 µg/ml of Hg(2+) in 2 to 5% HNO3
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only