Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Phosphorus standard solution, for AAS, 1 mg/ml P in water
Thermo Scientific Chemicals

Phosphorus standard solution, for AAS, 1 mg/ml P in water

CAS: 7723-14-0 | P | 30.97 g/mol
Have Questions?
Catalog NumberQuantity
196185000500 mL
Catalog number 196185000
Price (EUR)
60,65
Online exclusive
67,20
Save 6,55 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
60,65
Online exclusive
67,20
Save 6,55 (10%)
Each
Specifications
Assay Percent Range98%
CAS7723-14-0
CAS7732-18-5
ChEBICHEBI:30207
Chemical Name or MaterialPhosphorus standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only