Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl
Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl
Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl
Thermo Scientific Chemicals

Platinum standard solution, for AAS, 1 mg/ml Pt in 10-20% HCl

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
196191000100 mL
Catalog number 196191000
Price (USD)
80.50
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
80.50
Each
Specifications
CAS7440-06-4
CAS7647-01-0
CAS7732-18-5
ChEBICHEBI:33400
Chemical Name or MaterialPlatinum standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only