Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3
Thermo Scientific Chemicals

Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3

CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Catalog NumberQuantity
196205000500 mL
Catalog number 196205000
Price (USD)
108.65
Special offer
Online exclusive
Ends: 28-Dec-2026
128.00
Save 19.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
108.65
Special offer
Online exclusive
Ends: 28-Dec-2026
128.00
Save 19.35
Each
Specifications
CAS7440-09-7
CAS7732-18-5
ChEBICHEBI:26216
Chemical Name or MaterialPotassium standard solution
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only