Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals
Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals
Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhenium standard solution, for AAS, 1mg/mL Re in water, Thermo Scientific Chemicals

CAS: 13598-65-7 | Molecular Formula: H4NO4Re | Molecular Weight: 268.24 g/mol
Have Questions?
Catalog NumberQuantity
196221000100 mL
Catalog number 196221000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS13598-65-7
CAS7732-18-5
Chemical Name or MaterialRhenium standard solution
ColorColorless
IUPAC Namerheniumoylol amine
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy

RUO – Research Use Only