Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
Thermo Scientific Chemicals

Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl

CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 196281000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.30
Save 6.65
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.30
Save 6.65
Each
Specifications
CAS7782-49-2
CAS7647-01-0
CAS7732-18-5
ChEBICHEBI:27568
Chemical Name or MaterialSelenium standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only