Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH
Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH
Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH
Thermo Scientific Chemicals

Silicon standard solution, for AAS, 1 mg/ml Si in 2% NaOH

CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Catalog NumberQuantity
196291000100 mL
Catalog number 196291000
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.90
Save 15.25
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.90
Save 15.25
Each
Specifications
CAS7440-21-3
ChEBICHEBI:27573
Chemical Name or MaterialSilicon standard solution
Density1.0200g/mL
Formula Weight28.09
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopy
RUO – Research Use Only