Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
Thermo Scientific Chemicals

Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 196435000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS7440-62-2
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:35170
Chemical Name or MaterialVanadium standard solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only