Potassium bromide, 98%, extra pure
Potassium bromide, 98%, extra pure
Potassium bromide, 98%, extra pure
Thermo Scientific Chemicals

Potassium bromide, 98%, extra pure

CAS: 7758-02-3 | BrK | 119.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 196480050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=97.5 % (on dry substance)
Heavy metals (as Pb)=<10 ppm
Sulfate (SO4)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only