Zinc chloride, 98%, extra pure
Zinc chloride, 98%, extra pure
Zinc chloride, 98%, extra pure
Thermo Scientific Chemicals

Zinc chloride, 98%, extra pure

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 196840010
Price (USD)
139.00
Each
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
139.00
Each
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Complexometric97.5 to 102.5 %
Cadmium (Cd)=<10 ppm
Copper (Cu)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only