Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous
Thermo Scientific Chemicals

Magnesium chloride, 99.9%, (trace metal basis), pure, anhydrous

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Plastic bottle
Catalog number 197060100
Price (USD)
89.65
Special offer
Online exclusive
Ends: 28-Dec-2026
105.00
Save 15.35
Each
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
89.65
Special offer
Online exclusive
Ends: 28-Dec-2026
105.00
Save 15.35
Each
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.1 %
Appearance (Form)Powder to beads
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only