Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized
Thermo Scientific Chemicals

Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized

CAS: 20816-12-0 | O4Os | 254.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 197450010
Price (USD)
-
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20816-12-0
IUPAC Nametetraoxoosmium
Molecular FormulaO4Os
InChI KeyVUVGYHUDAICLFK-UHFFFAOYSA-N
SMILESO=[Os](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Fused mass or solution
Strength>=2.5 wt%
Stabilizertert-Butyl hydroperoxide
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only