Urea, 98%, extra pure, pearls
Urea, 98%, extra pure, pearls
Urea, 98%, extra pure, pearls
Thermo Scientific Chemicals

Urea, 98%, extra pure, pearls

CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 197460010
Price (USD)
72.65
Online exclusive
80.00
Save 7.35
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
72.65
Online exclusive
80.00
Save 7.35
Each
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.4 %
Impurity=<1.2 % (Biuret)
Appearance (Form)Pearls
Infrared spectrumConforms
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only