Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Thermo Scientific Chemicals

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh

CAS: 29998-99-0 | C8H16O10Rh2 | 478.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 197610010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS29998-99-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay43 % (Rh)
Appearance (Form)Powder
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only