Ammonium iron (III) citrate, 16.5-18.5% Fe, FCC, GMP, J.T.Baker™
Avantor™

Ammonium iron (III) citrate, 16.5-18.5% Fe, FCC, GMP, J.T.Baker™

Food GMP manufactured product, meets FCC specifications.
Have Questions?
Número do catálogo 1980-01
Preço (BRL)
-
Food GMP manufactured product, meets FCC specifications.
Food GMP manufactured product, meets FCC specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificações
Boiling Point197°C (1013 hPa)
CAS1185-57-5
Chemical Name or MaterialAmmonium iron (III) citrate
ColorBrown
Molecular FormulaC6H5+4(y)Fe(x)N(y)O7
PackagingAmber glass bottle, wide mouth, tamper-evident cap
SynonymAmmonium iron (III) citrate, Ferric Ammonium Citrate, Ammonium ferric citrate
GradeFCC
Molecular Weight (g/mol)264.99
Percent Purity16.5 to 18.5% Fe
Physical FormSolid
Quantity500 g
Unit SizeEach