Aluminum bromide, 98+%, extra pure, anhydrous
Aluminum bromide, 98+%, extra pure, anhydrous
Aluminum bromide, 98+%, extra pure, anhydrous
Thermo Scientific Chemicals

Aluminum bromide, 98+%, extra pure, anhydrous

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 198410250
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.80
Save 9.15
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 28-Dec-2026
60.80
Save 9.15
Each
Chemical Identifiers
CAS7727-15-3
IUPAC Namealuminium(3+) tribromide
Molecular FormulaAlBr3
InChI KeyPQLAYKMGZDUDLQ-UHFFFAOYSA-K
SMILES[Al+3].[Br-].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or chunks
Titration Complexometric>=98.0 %
Chloride (Cl)=<2000 ppm
Sulfate (SO4)=<200 ppm
Appearance (Color)White to yellow to pink to grey
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only