Can I purchase the individual components of the ViewRNA Tissue Assay Core Kit and ViewRNA Tissue Assay Blue Module as standalone products?
Yes. Please refer to the catalog numbers listed on pages 4 and 5 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018633_viewRNA_ISH_UG.pdf).