Search
View additional product information for ViewRNA™ Tissue Assay Blue Module - FAQs (19932, 19943)
4 product FAQs found
Yes. Please refer to the catalog numbers listed on pages 4 and 5 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018633_viewRNA_ISH_UG.pdf).
Yes. It is offered separately as Label Probe Diluent QF, Cat no. QVT0513.
Yes. There was no change to the Fast Blue reagent used.
The earlier Fast Blue Substrate products (Cat Nos. QVT0506 and QVT0507) provided only the Blue Buffer and Blue Reagents 1, 2, and 3. The ViewRNA Tissue Assay Blue Module (Cat Nos. 19932, 19943) includes these same components plus a Label Probe 6-AP and a stop reagent.