Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH
Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH
Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, extra pure, ca. 56% LiOH

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 199540050
Price (USD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl>=54 % (LiOH)
Insoluble matter=<0.1 % (in water)
Sodium + Potassium (Na+K)=<3000 ppm
Carbon dioxide (CO2)=<3500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only