Acetato de litio dihidrato, 98 %, extra puro, Thermo Scientific Chemicals
Acetato de litio dihidrato, 98 %, extra puro, Thermo Scientific Chemicals
Acetato de litio dihidrato, 98 %, extra puro, Thermo Scientific Chemicals
Acetato de litio dihidrato, 98 %, extra puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetato de litio dihidrato, 98 %, extra puro, Thermo Scientific Chemicals

CAS: 6108-17-4 | C2H3LiO2·2H2O | 102.02 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1 kg
Envase:
Frasco de plástico
Número de catálogo 199840010
Precio (USD)
-
Cantidad:
1 kg
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS6108-17-4
IUPAC Namelithium(1+) acetate dihydrate
Molecular FormulaC2H7LiO4
InChI KeyIAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES[Li+].O.O.CC([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
pH7.5 to 9.5 (5 % solution)
Sodium (Na)=<50 ppm
Appearance (Form)Adhering crystalline powder and/or chunks
Insoluble matter=<0.01 % (in water)
Titration with HClO4>=97.5 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only