1,2-Octanediol, 98+%
1,2-Octanediol, 98+%
1,2-Octanediol, 98+%
Thermo Scientific Chemicals

1,2-Octanediol, 98+%

CAS: 1117-86-8 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 200140100
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.60
Save 4.95
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 28-Dec-2026
31.60
Save 4.95
Each
Chemical Identifiers
CAS1117-86-8
IUPAC Nameoctane-1,2-diol
Molecular FormulaC8H18O2
InChI KeyAEIJTFQOBWATKX-UHFFFAOYNA-N
SMILESCCCCCCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only