Diethylcyanamide, 99+%
Diethylcyanamide, 99+%
Diethylcyanamide, 99+%
Thermo Scientific Chemicals

Diethylcyanamide, 99+%

CAS: 617-83-4 | C5H10N2 | 98.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 200220500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS617-83-4
IUPAC Namecyanodiethylamine
Molecular FormulaC5H10N2
InChI KeyZZTSQZQUWBFTAT-UHFFFAOYSA-N
SMILESCCN(CC)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4210 to 1.4240 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only