Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles
Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles
Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles
Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles
Thermo Scientific™

Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles

Store, ship and package light-sensitive liquids using these narrow-mouth, amber HDPE bottles that offer excellent chemical resistance to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2004-0002PK60 mL20 mmPack of 12
2004-91254 mL13 mmCase of 72
2004-9125PK
also known as 2004-9125
4 mL13 mmPack of 12
2004-90258 mL20 mmCase of 72
2004-9025PK
also known as 2004-9025
8 mL20 mmPack of 12
2004-905015 mL20 mmCase of 72
2004-9050PK
also known as 2004-9050
15 mL20 mmPack of 12
2004-000130 mL20 mmCase of 72
2004-0001PK
also known as 2004-0001
30 mL20 mmPack of 12
2004-000260 mL20 mmCase of 72
2004-0004125 mL24 mmCase of 72
2004-0004PK
also known as 2004-0004
125 mL24 mmPack of 12
2004-0008250 mL24 mmCase of 72
2004-0008PK
also known as 2004-0008
250 mL24 mmPack of 12
2004-0016500 mL28 mmCase of 48
2004-0016PK
also known as 2004-0016
500 mL28 mmPack of 12
2004-00321 L38-430 mmCase of 24
2004-0032PK
also known as 2004-0032
1 L38-430 mmPack of 6
Catalog number 2004-0002PK
Price (USD)
42.71
Online exclusive
47.46
Save 4.75
Pack of 12
Capacity (Metric):
60 mL
Closure Size:
20 mm
Unit Size:
Pack of 12
Price (USD)
42.71
Online exclusive
47.46
Save 4.75
Pack of 12

Store, ship and package light-sensitive liquids using Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Lab Quality Bottles. Featuring excellent chemical resistance to most acids, bases and alcohols, these high-density polyethylene bottles are leakproof† when used with appropriate Nalgene closures. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.

Features:
  • Opaque amber reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 requirements for light transmission
  • Versatile bottles where exposure to light needs to be restricted
  • Can be used in freezer to -100°C (-148°F) for long term storage
  • Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaiminate your reagents
  • Leakproof† when used with Nalgene closures

Includes:

Amber polypropylene screw caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.



Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity2 oz., 60 mL
Capacity (English)2 oz.
Certifications/ComplianceMeet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure ColorAmber
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size20 mm
Closure TypeScrew Cap
DescriptionLab Quality Amber Narrow-Mouth Bottle
Diameter (Metric)37 mm
Diameter (Metric) Inner Neck14 mm
Height (English)3.36 in.
Height (Metric)85 mm
Neck Finish20 mm
No. per Case72
No. per Pack12
Product TypeStorage Bottle
ShapeRound
Through the Closure DispensingNo
Width (English)1.52 in.
Capacity (Metric)60 mL
Closure IncludedYes
ColorOpaque Amber
GraduatedNo
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizePack of 12

Frequently asked questions (FAQs)

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.