ART™ Pipette Tip Empty Hinged Racks
Thermo Scientific™
ART™ Pipette Tip Empty Hinged Racks
Save valuable space in the lab with these stackable pipette tip hinged racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeUnit Size
2006-ERMediumCase of 50
2004-ERSmallCase of 50
2004-ERPK
also known as 2004-ER
SmallPack of 10
2006-ERPK
also known as 2006-ER
MediumPack of 10
2008-ERLargeCase of 32
2008-ERPK
also known as 2008-ER
LargePack of 8
Catalog number 2006-ER
Price (USD)
164.85
Online exclusive
235.50
Save 70.65
Case of 50
Size:
Medium
Unit Size:
Case of 50
Price (USD)
164.85
Online exclusive
235.50
Save 70.65
Case of 50
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ Pipette Tip Empty Hinged Racks. ART barrier tips provide 100% security against aerosols and liquid contamination. Front latch closes and locks, ensuring safe and secure storage throughout.
Features:
  • ART barrier tips provide 100% security against aerosols and liquid contamination
  • Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
Certifications:
  • Nonsterile products certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Sterile products certified to be free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Certifications/ComplianceCertified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
QuantityHinged Rack, 10 trays/Pk., 5 Pks,/Cs.
SizeMedium
Product TypeEmpty Hinged Rack
ShapeSquare
StackableYes
SterilityNon-sterile
Unit SizeCase of 50