Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Nalgene™ Rectangular Amber HDPE Bottles
Thermo Scientific™

Nalgene™ Rectangular Amber HDPE Bottles

Protect light-sensitive liquids or dry materials from exposure with these space-efficient, amber HDPE bottles, featuring sturdy walls and a wide mouth for easy filling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2009-0008250 mL38 mmCase of 72
2009-0004125 mL28 mmCase of 72
2009-0004PK
also known as 2009-0004
125 mL28 mmPack of 12
2009-0008PK
also known as 2009-0008
250 mL38 mmPack of 12
2009-0016500 mL48 mmCase of 48
2009-0016PK
also known as 2009-0016
500 mL48 mmPack of 12
2009-00321 L53 mmCase of 24
2009-0032PK
also known as 2009-0032
1 L53 mmPack of 6
2009-00642 L63 mmCase of 12
2009-0064PK
also known as 2009-0064
2 L63 mmPack of 4
Catalog number 2009-0008
Price (USD)
416.46
Online exclusive
489.95
Save 73.49
Case of 72
Capacity (Metric):
250 mL
Closure Size:
38 mm
Unit Size:
Case of 72
Price (USD)
416.46
Online exclusive
489.95
Save 73.49
Case of 72

Protect light-sensitive liquids or dry materials from exposure. Thermo Scientific™ Nalgene™ Rectangular Amber HDPE Bottles take up less shelf space than traditional round bottles. These high-density, polyethylene bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Excellent chemical resistance to most acids, bases and alcohols makes this container great for sampling, shipping or storing liquids or dry materials that are sensitive to UV light.

  • Amber polypropylene screw closure
  • Amber HDPE reduces UV light transmissions to protect light-sensitive liquids
  • Meets U.S. Pharmacopoeia 23 light transmission requirements
  • Rectangular shape is great for space-saving use in the freezer to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
.

Recycling Information
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity8 oz., 250 mL
Capacity (English)8 oz.
Certifications/ComplianceMeet U.S. Pharmacopeia XXIII requirements for maximum light transmission.
Closure ColorOpaque Amber
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38 mm
Closure TypeScrew Cap
DescriptionAmber Wide-Mouth Rectangular Bottle
Diameter (English) Inner Neck1.1 in.
Diameter (Metric)76.2 mm
Diameter (Metric) Inner Neck28 mm
Height (English)4.6 in.
Height (Metric)117 mm
Length (Metric)76 mm
No. per Case72
No. per Pack12
Product TypeWide Mouth Bottle
ShapeRectangular
Through the Closure DispensingNo
Width (Metric)51 mm
Capacity (Metric)250 mL
Closure IncludedYes
ColorAmber
GraduatedNo
MaterialHDPE
MouthWide
SterilityNon-sterile
Unit SizeCase of 72

Frequently asked questions (FAQs)

Are your Nalgene containers tested for air shipment of liquid samples?

Most of our products are manufactured and tested for leakproof performance in laboratory and industrial settings only. Our PassPort IP2 bottles (2099 and 2199 series) represent the only exception to this: they are rigorously leak tested under a 15 psi (-95 kPa) pressure differential to simulate unpressurized air cargo transport.

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.