Matrix™ WellMate™ Disposable Tubing Cartridges, Small Bore - FAQs
Matrix™ WellMate™ Disposable Tubing Cartridges, Small Bore - FAQs
View additional product information for Matrix™ WellMate™ Disposable Tubing Cartridges - FAQs (201-30002)
2 product FAQs found
Do Matrix WellMate disposable tubing cassettes have an expiration date?
We do not provide shelf life or expiration dates for the cassettes because they are a consumable not meant to be stored for long periods before use. Typically, cassettes are used within a year's time after purchase. When not in use, we recommend that you store the cassettes in their original packaging, away from extreme temperatures or conditions that would cause damage to the materials. For optimum performance, we recommend storing the cassettes at 5 - 40 degrees C.
Do Matrix WellMate disposable tubing cassettes come with a Certificate of Analysis (COA)?
All WellMate dispensing cassettes are calibrated individually, and the calibration report is included with each dispensing cassette. The Calibration Certificate/Factory Calibration Report provided for filing under GLM/GLP is also referred to as the Certificate of Analysis (COA). The COA contains pertinent information such as the catalog number, serial number, and production lot number.