N,N-Dimethylmethyleneiminium iodide, 97%
N,N-Dimethylmethyleneiminium iodide, 97%
N,N-Dimethylmethyleneiminium iodide, 97%
Thermo Scientific Chemicals

N,N-Dimethylmethyleneiminium iodide, 97%

CAS: 33797-51-2 | C3H8IN | 185.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 201010100
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS33797-51-2
IUPAC Name(dimethylamino)methylium iodide
Molecular FormulaC3H8IN
InChI KeyVVDUZZGYBOWDSQ-UHFFFAOYSA-M
SMILES[I-].CN(C)[CH2+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to orange to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only