Sulfur, 99.5%, sublimed
Sulfur, 99.5%, sublimed
Sulfur, 99.5%, sublimed
Thermo Scientific Chemicals

Sulfur, 99.5%, sublimed

CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 201250250
Price (USD)
45.65
Online Exclusive
50.30
Save 4.65 (9%)
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
45.65
Online Exclusive
50.30
Save 4.65 (9%)
Each
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay>=99.4 %
Appearance (Form)Powder
Sulfated ash=<0.2 %
Free acid (H2SO4)=<0.02 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only