Acenaphthene, 99%
Acenaphthene, 99%
Acenaphthene, 99%
Thermo Scientific Chemicals

Acenaphthene, 99%

CAS: 83-32-9 | C12H10 | 154.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number 201340050
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.10
Save 5.45
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.10
Save 5.45
Each
Chemical Identifiers
CAS83-32-9
IUPAC Name1,2-dihydroacenaphthylene
Molecular FormulaC12H10
InChI KeyCWRYPZZKDGJXCA-UHFFFAOYSA-N
SMILESC1CC2=C3C1=CC=CC3=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point91°C to 95°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only