Ammonium cerium(IV) nitrate, 99%, for analysis
Ammonium cerium(IV) nitrate, 99%, for analysis
Ammonium cerium(IV) nitrate, 99%, for analysis
Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, 99%, for analysis

CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 201441000
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
107.00
Save 16.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
90.65
Special offer
Online exclusive
Ends: 28-Dec-2026
107.00
Save 16.35
Each
Chemical Identifiers
CAS16774-21-3
IUPAC Nameλ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular FormulaCeH8N8O18
InChI KeyWIBGOERAEYJBOT-UHFFFAOYSA-N
SMILESN.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Crystalline powder or crystals
Titration>=98.5 % (on dry substance)
Iron(III)oxide (Fe2O3)=<5 ppm
Calcium oxide (CaO)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only