Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure, 1000mL - FAQs
View additional product information for Nalgene™ Square Narrow-Mouth PPCO Bottles with Closure - FAQs (2016-0250PK, 2016-0500PK, 2016-0500, 2016-0125, 2016-0030PK, 2016-1000, 2016-0060PK, 2016-0060, 2016-0030, 2016-0125PK, 2016-0250, 2016-1000PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.