Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Thermo Scientific™

Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2018-10001 L38-430Case of 24
2018-003030 mL20 mmCase of 72
2018-0030PK
also known as 2018-0030
30 mL20 mmPack of 12
2018-006060 mL24 mmCase of 72
2018-0060PK
also known as 2018-0060
60 mL24 mmPack of 12
2018-0125125 mL38-430Case of 72
2018-0125PK
also known as 2018-0125
125 mL38-430Pack of 12
2018-0250250 mL38-430Case of 72
2018-0250PK
also known as 2018-0250
250 mL38-430Pack of 12
2018-1000PK
also known as 2018-1000
1 L38-430Pack of 6
Catalog number 2018-1000
Price (USD)
235.24
Online exclusive
276.75
Save 41.51
Case of 24
Capacity (Metric):
1 L
Closure Size:
38-430
Unit Size:
Case of 24
Price (USD)
235.24
Online exclusive
276.75
Save 41.51
Case of 24

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof† when used with Nalgene closures
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
ACT LabelYes
AutoclavableNot Autoclavable
Capacity32 oz., 1000 mL
Capacity (English)32 oz.
Certifications/ComplianceUSP Class VI, FDA - 21 CFR
Closure ColorNatural, translucent
Closure MaterialPolypropylene
Closure Size38-430
Closure TypeScrew Cap
DescriptionNarrow-Mouth Square Bottle
Diameter (English)3.7 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric)92 mm
Diameter (Metric) Inner Neck28 mm
GraduationsYes
Height (English)8.6 in.
Height (Metric)218 mm
IncludesLeakproof polypropylene screw closures
No. per Case24
No. per Pack6
Product TypeSquare Narrow Mouth Bottle
ShapeSquare
Through the Closure DispensingNo
Width (English)3.9 in.
Capacity (Metric)1 L
Closure IncludedYes
ColorTranslucent
GraduatedYes
MaterialHDPE
MouthNarrow
SterilityNon-sterile
Unit SizeCase of 24