Nalgene™ Square PETG Media Bottles with Closure, 500 mL - FAQs
Nalgene™ Square PETG Media Bottles with Closure, 500 mL - FAQs
View additional product information for Nalgene™ Square PETG Media Bottles with Closure - FAQs (2019-0500, 2019-0125, 2019-0030, 2019-0250, 2019-2000, 2019-1000, 2019-0060)
2 product FAQs found
Are Nalgene Square PETG Media Bottles with Closure suitable for making aliquots of cell culture media?
Nalgene Square PETG Media Bottles with Closure can be used for cell culture media as they are sterile and non-pyrogenic.
Note: Make sure everything is done aseptically.
How accurate are the molded or printed graduations on your bottles, beakers, carboys and tanks?
The accuracy of these graduations cannot be guaranteed. We suggest that they not be relied upon for measurement, but rather to be used for reference only.