Nalgene™ Square PETG Media Bottles with Closure, 1 L - FAQs
Nalgene™ Square PETG Media Bottles with Closure, 1 L - FAQs
View additional product information for Nalgene™ Square PETG Media Bottles with Closure - FAQs (2019-0500, 2019-0125, 2019-0030, 2019-0250, 2019-2000, 2019-1000, 2019-0060)
3 product FAQs found
What is the difference between the following Nalgene Square PETG Media Bottles: Cat. No. 2019-xxxx (with Closure), Cat. No. 342020-xxxx (with Closure: Sterile, Shrink-Wrapped Trays), Cat. No. CE-N2019-xxxx (Platinum Certified Clean)?
All three products use the same PETG bottles, so the materials and specifications are the same for the bottle itself. Cat. No. 342020-xxxx is supplied with natural-colored HDPE closures, while Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx are supplied with white HDPE closures. The HDPE resin used for the closures is the same, but a white colorant has been added to the resin for Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx (this will be noted on the COA). Cat. No. 2019-xxxx also has a PVC tamper-resistant seal around the closure, while Cat. No. 342020-xxxx does not.
For more detail about the different "levels of clean" for our Thermo Scientific Nalgene contains, please refer to our Nalgene Clean Containers brochure.
Are Nalgene Square PETG Media Bottles with Closure suitable for making aliquots of cell culture media?
Nalgene Square PETG Media Bottles with Closure can be used for cell culture media as they are sterile and non-pyrogenic.
Note: Make sure everything is done aseptically.
How accurate are the molded or printed graduations on your bottles, beakers, carboys and tanks?
The accuracy of these graduations cannot be guaranteed. We suggest that they not be relied upon for measurement, but rather to be used for reference only.