Nalgene™ Square PETG Media Bottles with Closure
Thermo Scientific™

Nalgene™ Square PETG Media Bottles with Closure

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure, ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2019-20002000 mL
2019-003030 mL
2019-006060 mL
2019-0125125 mL
2019-0250250 mL
2019-0500500 mL
2019-10001 L
Catalog number 2019-2000
Price (MXN)
-
Capacity (Metric):
2000 mL

Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.

Features:

  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
  • Suitable for frozen storage down to -40°C
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Sterile and non-pyrogenic
  • Available in low particulate format
  • Supported by extensive validation documentation

Ordering Information:

  • Packed in shrink-wrapped trays

Note:

  • 2 L size has molded-in handgrips

Warranty:

  • 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AutoclavableNot Autoclavable
Capacity (English)64 oz.
Certifications/ComplianceBottles and closures are certified radiation sterilized (SAL 10-6), non-pyrogenic and non-cytotoxic, and comply with USP Class VI guidelines.
Closure MaterialHDPE
Closure Size53B
Diameter (English)4.57 in.
Diameter (English) Inner Neck1.5 in.
Diameter (Metric)116 mm
Diameter (Metric) Inner Neck39 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage
Height (English)10.67 in.
Height (Metric)271 mm
No. per Case12/Cs.
No. per Pack6/Pk.
Product TypeMedia Bottle
ShapeSquare
Temperature Range (Metric)-70°C to 70°C
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage. Upstream Validation
Capacity (Metric)2000 mL
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPETG
MouthNarrow
SterilitySterile
Unit SizeCase of 12

Frequently asked questions (FAQs)

What is the difference between the following Nalgene Square PETG Media Bottles: Cat. No. 2019-xxxx (with Closure), Cat. No. 342020-xxxx (with Closure: Sterile, Shrink-Wrapped Trays), Cat. No. CE-N2019-xxxx (Platinum Certified Clean)?

All three products use the same PETG bottles, so the materials and specifications are the same for the bottle itself. Cat. No. 342020-xxxx is supplied with natural-colored HDPE closures, while Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx are supplied with white HDPE closures. The HDPE resin used for the closures is the same, but a white colorant has been added to the resin for Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx (this will be noted on the COA). Cat. No. 2019-xxxx also has a PVC tamper-resistant seal around the closure, while Cat. No. 342020-xxxx does not.

For more detail about the different "levels of clean" for our Thermo Scientific Nalgene contains, please refer to our Nalgene Clean Containers brochure.

Are Nalgene Square PETG Media Bottles with Closure suitable for making aliquots of cell culture media?

Nalgene Square PETG Media Bottles with Closure can be used for cell culture media as they are sterile and non-pyrogenic.
Note: Make sure everything is done aseptically.

How accurate are the molded or printed graduations on your bottles, beakers, carboys and tanks?

The accuracy of these graduations cannot be guaranteed. We suggest that they not be relied upon for measurement, but rather to be used for reference only.